Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Paxillin Antibody (LD4 sAB), FITC, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP281063F
Description
Paxillin Monoclonal specifically detects Paxillin in Human samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.Specifications
Paxillin | |
Monoclonal | |
FITC | |
PBS | |
PXN | |
Peptide corresponding to human paxillin-LD4 motif (residues 260-278) fused to MBP; peptide sequence: WGGSATRELDELMASLSD. | |
0.1 mL | |
Cell Biology, Cellular Markers, Phospho Specific, Signal Transduction | |
5829 | |
Human | |
Purified |
ELISA, Immunocytochemistry, Immunofluorescence | |
LD4 sAB | |
ELISA, Immunocytochemistry/Immunofluorescence | |
FLJ16691, paxillin | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
The antibody binds specifically to the LD-4 motif of human Paxillin with a Kd of 6 nM. | |
Store at 4°C in the dark. | |
IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction