Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PAXIP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543699
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543699 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543699 Supplier Invitrogen™ Supplier No. PA543699
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: MFDDSSDSSP EKQERNLNWT PAEVPQLAAA KRRLPQGKEP GLINLCANVP Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 79%.

This gene is a member of the paired box gene family and encodes a nuclear protein with six BRCT domains. This protein plays a critical role in maintaining genome stability, condensation of chromatin and progression through mitosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PAXIP1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene PAXIP1
Gene Accession No. Q6ZW49
Gene Alias CAGF28; CAGF29; D5Ertd149e; fa04b06; FLJ41049; PACIP1; PAX interacting (with transcription-activation domain) protein 1; PAX interacting protein 1; PAX transactivation activation domain-interacting protein; PAX transcription activation domain interacting protein 1 like; PAX-interacting protein 1; PAX-interacting protein 1-like protein; Paxip1; PAXIP1L; protein encoded by CAG trinucleotide repeats; PTIP; TNRC2; wu:fa04b06; zgc:92593
Gene Symbols PAXIP1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human PAXIP1 (aa 248-297).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22976
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.