Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PAXIP1 Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™

Catalog No. ABE187725UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABE1877-25UL 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABE1877-25UL Supplier MilliporeSigma Supplier No. ABE187725UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-PAXIP1 is a highly specific rabbit polyclonal antibody, that targets PAX-interacting protein 1 and has been tested in Western Blotting, Chromatin Immunoprecipitation (ChIP), ChIP-seq and Immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PAXIP1
Applications ChIP Assay, Immunoprecipitation
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols Paxip1,Ptip
Host Species Rabbit
Immunogen His-tagged recombinant fragment corresponding to 199 amino acids from the N-terminal region of murine PAX-interacting protein 1.
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Test Specificity This rabbit polyclonal antibody detects PAX-interacting protein 1 in human and murine cells. It targets an epitope within 199 amino acids from the N-terminal region.
Target Species Human, Mouse
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.