Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PAXX Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 2 publications
Supplier: Novus Biologicals NBP19417225UL
Description
PAXX Polyclonal specifically detects PAXX in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockout Validated, Knockdown Validated.Specifications
C9orf142 | |
Polyclonal | |
Western Blot 0.04-0.4 ug/ml, Immunohistochemistry 1:2500 - 1:5000, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:2500 - 1:5000, Knockout Validated, Knockdown Validated | |
chromosome 9 open reading frame 142, hypothetical protein LOC286257 | |
Rabbit | |
Affinity Purified | |
RUO | |
286257 | |
Human, Mouse | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoassay, KnockDown | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
C9orf142 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:LSKVPGPEAAPRLRALTLGLAKRVWSLERRLAAAEETAVSPRKSPRPAGPQLFLPDPDPQRGGPGPGVRRRCPGESLINPGFKSKKPAGGVDFDET | |
25 μL | |
Primary | |
Specificity of human PAXX antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction