Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PAXX, Rabbit anti-Human, Polyclonal, Millipore Sigma™

Catalog No. ABE179525UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABE179525UG 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABE179525UG Supplier MilliporeSigma Supplier No. ABE179525UG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-PAXX, Cat. No. ABE1795, is a highly specific rabbit polyclonal antibody that targets PAXX (C9orf142) and has been tested for use in Immunohistochemistry (Paraffin) and Western Blotting.

Protein PAXX (UniProt: 9BUH6; also known as Paralog of XRCC4 and XLF, XRCC4-like small protein) is encoded by the C9orf142 (also known as PAXX, XLS) gene (Gene ID: 286257) in human. PAXX is a newer member of XRCC4 superfamily that predominantly localizes to the nucleus and is shown to accumulate at sites of DNA damage generated by laser microirradiation. It is involved in non-homologous end joining (NHEJ), a major pathway to repair double-strand breaks in DNA and may act as a scaffold required to stabilize the Ku heterodimer, composed of XRCC5/Ku80 and XRCC6/Ku70, at double-strand break sites where it promote the assembly and/or stability of the NHEJ machinery. PAXX residues 1-113 form a head domain, which is structurally related to those of XRCC4, XLF and SAS6. This is followed by a 31-amino-acid -helix, forming a coiled-coil with the other protomer to make a PAXX homodimer. Two isoforms of PAXX have been described that are produced by alternative splicing. (Ref.: Ochi, T., et al. (2015). Science 347(6218); 185-188).
TRUSTED_SUSTAINABILITY

Specifications

Antigen PAXX
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry Analysis: A 1:1,000 dilution from a representative lot detected PAXX in human prostate, human tonsil, and human cervical cancer tissues.
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1M Tris-Glycine, 0.15M NaCl, pH7.4 with 0.05% sodium azide.
Gene Alias Protein PAXX;Paralog of XRCC4 and XLF;XRCC4-like small protein;C9orf142
Gene Symbols C9orf142;PAXX;XLS
Host Species Rabbit
Immunogen KLH-conjugated liner peptide corresponding to 16 amino acids from the C-terminus region of human Protein PAXX.
Molecular Weight of Antigen ∽22 kDa observed; 21.64 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Epigenetics & Nuclear Function
Primary or Secondary Primary
Test Specificity This rabbit polyclonal antibody detects PAXX protein. It targets an epitope within 16 amino acids from the C-terminus region.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.