Learn More
Tocris Bioscience™ PB 28 dihydrochloride

Description
Specifications
Specifications
| Chemical Name or Material | 1-Cyclohexyl-4-[3-(1,2,3,4-tetrahydro-5-methoxy-1-naphthalenyl)propyl]piperazine dihydrochloride |
| CAS | 172907-03-8 |
| Assay Percent Range | >99% |
| Quantity | 10 mg |
| Solubility | Soluble to 20mM with gentle warming in water |
| Molecular Formula | C24H38N2O.2HCl |
| Formula Weight | Observed MW: 443.5 |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.