Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PBDC1 Antibody, Alexa Fluor™ 750, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP318459AF750
Description
PBDC1 Polyclonal antibody specifically detects PBDC1 in Human samples. It is validated for Western BlotSpecifications
PBDC1 | |
Polyclonal | |
Western Blot | |
chromosome X open reading frame 26, hypothetical protein LOC51260, MGC874,2610029G23Rik | |
Recognizes region between residues Between Between 183 and 233.. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
Alexa Fluor 750 | |
50mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
51260 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction