Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PBDC1 Antibody (OTI4E11), PE/Atto594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271912PEATT594
Description
PBDC1 Monoclonal specifically detects PBDC1 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.Specifications
| PBDC1 | |
| Monoclonal | |
| PE/ATTO 594 | |
| PBS with 0.05% Sodium Azide | |
| PBDC1 | |
| Full length recombinant protein of human CXorf26 (NP_057584) produced in HEK293T cells. | |
| 0.1 mL | |
| 51260.0 | |
| Store at 4°C in the dark. Do not freeze. |
| Flow Cytometry | |
| OTI4E11 | |
| Flow Cytometry | |
| chromosome X open reading frame 26, hypothetical protein LOC51260, MGC874,2610029G23Rik | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction