Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PBDC1 Antibody (OTI5A11) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271911
Description
PBDC1 Monoclonal specifically detects PBDC1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
CXorf26 | |
Monoclonal | |
LYOPH | |
Western Blot 1:2000, Immunohistochemistry 1:50-150 | |
chromosome X open reading frame 26, hypothetical protein LOC51260, MGC874,2610029G23Rik | |
Mouse | |
25.9 kDa | |
100 μg | |
51260 | |
Human, Mouse, Rat | |
Lyophilized |
Western Blot, Immunohistochemistry | |
OTI5A11 | |
Unconjugated | |
Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
PBDC1 | |
Full length human recombinant protein of human CXorf26(NP_057584) produced in HEK293T cell. | |
Affinity Purified | |
Primary | |
Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction