Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PBDC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551842
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551842 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551842 Supplier Invitrogen™ Supplier No. PA551842
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EVYYKLISSV DPQFLKLTKV DDQIYSEFRK NFETLRIDVL DPEELKSESA KEKWRPFCLK FNGIVEDFNY GTLLRLDCSQ GYTEENTIFA PRIQFFAIEI ARNREGYNKA VYISVQDKEG EKGVNNGGEK RADSGEEENT KNGGEK Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 85%.

PBDC1 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PBDC1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Pbdc1
Gene Accession No. Q9BVG4
Gene Alias 2610029G23Rik; CXorf26; MGC874; Pbdc1; polysaccharide biosynthesis domain containing 1; polysaccharide biosynthesis domain-containing protein 1; Protein PBDC1; UPF0368 protein Cxorf26; UPF0368 protein Cxorf26 homolog
Gene Symbols Pbdc1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PBDC1. Recombinant protein control fragment (Product #RP-92192).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51260
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.