Learn More
PBDC1 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A305686AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
PBDC1 is a protein coding gene.Specifications
PBDC1 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q9BVG4 | |
PBDC1 | |
Between 1 and 50 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunoprecipitation, Western Blot | |
Unconjugated | |
PBDC1 | |
2610029G23Rik, chromosome X open reading frame 26, CXorf26, MGC874, polysaccharide biosynthesis domain-containing protein 1, UPF0368 protein Cxorf26 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51260 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.