Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PBEF/Visfatin/NAMPT Antibody (362616), Alexa Fluor™ 700, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB4044N
Description
PBEF/Visfatin/NAMPT Monoclonal antibody specifically detects PBEF/Visfatin/NAMPT in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOFSpecifications
PBEF/Visfatin/NAMPT | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Western Blot, Flow Cytometry, CyTOF | |
362616 | |
Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
EC 2.4.2.12, MGC117256, NAmPRTase, Nampt, nicotinamide phosphoribosyltransferase, PBEF1110035O14Rik, PBEF1DKFZp666B131, Pre-B cell-enhancing factor, pre-B-cell colony enhancing factor 1, Pre-B-cell colony-enhancing factor 1, VF, VISFATIN | |
Mouse myeloma cell line NS0-derived recombinant mouse PBEF/Visfatin, Met1-His491, Accession # Q99KQ4 | |
0.1 mL | |
Adaptive Immunity, Cell Cycle and Replication, Immunology, Innate Immunity | |
10135 | |
Store at 4C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction