Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PBK Antibody, Alexa Fluor™ 532, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299521AF532
Description
PBK Polyclonal specifically detects PBK in Human samples. It is validated for ELISA.Specifications
PBK | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
Alexa Fluor 532 | |
Cancer/testis antigen 84, CT84MAPKK-like protein kinase, EC 2.7.12.2, FLJ14385, Nori-3PDZ-binding kinase, PDZ binding kinase, serine/threonine protein kinase, Spermatogenesis-related protein kinase, SPKT-LAK cell-originated protein kinase, TOPKlymphokine-activated killer T-cell-originated protein kinase | |
Produced in rabbits immunized with purified, recombinant PBK (NP_060962.2; Met 1-Val 322). | |
0.1 mL | |
Cell Cycle and Replication | |
55872 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction