Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PBK Monoclonal Antibody (10)
GREENER_CHOICE

Catalog No. PIMA530729
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA530729 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA530729 Supplier Invitrogen™ Supplier No. MA530729
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human PBK/TOPK.

PBK (TOPK) is a serine/threonine kinase that is related to the dual specific mitogen-activated protein kinase (MAPKK). The kinase is up-regulated in a variety of hematological malignancies, participates in activating lymphoid cells, and supports testicular function (spermatogenesis). Evidence suggests that mitotic phosphorylation is required for its catalytic activity. This mitotic kinase may be involved in the activation of lymphoid cells and support testicular functions, with a suggested role in the process of spermatogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PBK
Applications ELISA
Classification Monoclonal
Clone 10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene PBK
Gene Accession No. Q96KB5
Gene Alias 2810434B10Rik; AW538537; Cancer/testis antigen 84; CT84; D14Ertd732e; epididymis luminal protein 164; HEL164; Lymphokine-activated killer T-cell-originated protein kinase; MAPKK-like protein kinase; Nori-3; PBK; PDZ binding kinase; PDZ-binding kinase; serine/threonine protein kinase; spermatogenesis-related protein kinase; SPK; T-LAK cell-originated protein kinase; TOPK
Gene Symbols PBK
Host Species Mouse
Immunogen Recombinant Human PBK/TOPK Protein (Met1-Val322).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55872
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.