Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PBLD Antibody (OTI7G5), PE/Cy7, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273262PECY7
Description
PBLD Monoclonal specifically detects PBLD in Human samples. It is validated for Flow Cytometry.Specifications
PBLD | |
Monoclonal | |
PE-Cyanine7 | |
PBS with 0.05% Sodium Azide | |
PBLD | |
Full length recombinant protein of human PBLD (NP_071412) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Flow Cytometry | |
OTI7G5 | |
Flow Cytometry | |
EC 5.1, FLJ14767, FLJ35507, MAWBPMAWDBPMAWD binding protein, MAWD-binding protein, phenazine biosynthesis-like domain-containing protein, phenazine biosynthesis-like protein domain containing, Unknown protein 32 from 2D-page of liver tissue | |
Mouse | |
Affinity Purified | |
Proteases & Other Enzymes | |
64081.0 | |
Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction