Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PBLD Monoclonal Antibody (OTI2B5), TrueMAB™, OriGene
SDP

Catalog No. 502541990
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-1990 30 μL
50-167-2934 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-1990 Supplier OriGene Supplier No. TA501824S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PBLD Monoclonal antibody specifically detects PBLD in Canine, Human samples. It is validated for Flow Cytometry, Western Blot

Specifications

Antigen PBLD
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone OTI2B5
Concentration 0.38 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene PUS7
Gene Accession No. Q96PZ0
Gene Alias 0610038K03Rik; epididymis secretory protein Li 306; HEL-S-306; Mawbp; Mawbp2; MAWD binding protein; MAWD binding protein homolog 1; MAWD-binding protein; MAWDBP; Pbld; Pbld1; phenazine biosynthesis like protein domain containing; phenazine biosynthesis-like domain-containing protein; Phenazine biosynthesis-like domain-containing protein 1; phenazine biosynthesis-like protein domain containing; phenazine biosynthesis-like protein domain containing 1; similar to MAWD binding protein (Unknown protein 32 from 2D-page of liver tissue); sphenazine biosynthesis-like protein domain containing; Unknown protein 32 from 2D-page of liver tissue
Host Species Mouse
Immunogen Full length human recombinant protein of human PUS7 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100856353, 54517
Target Species Canine, Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.