Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PBLD Monoclonal Antibody (OTI7G5)
GREENER_CHOICE

Catalog No. PIMA525311
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA525311 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA525311 Supplier Invitrogen™ Supplier No. MA525311
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PBLD Monoclonal Antibody for Western Blot, ICC/IF, Flow

The function of the PBLD protein remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PBLD
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone OTI7G5
Concentration 0.87 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PBLD
Gene Accession No. P30039
Gene Alias 0610038K03Rik; epididymis secretory protein Li 306; HEL-S-306; Mawbp; Mawbp2; MAWD binding protein; MAWD binding protein homolog 1; MAWD-binding protein; MAWDBP; Pbld; Pbld1; phenazine biosynthesis like protein domain containing; phenazine biosynthesis-like domain-containing protein; Phenazine biosynthesis-like domain-containing protein 1; phenazine biosynthesis-like protein domain containing; phenazine biosynthesis-like protein domain containing 1; similar to MAWD binding protein (Unknown protein 32 from 2D-page of liver tissue); sphenazine biosynthesis-like protein domain containing; Unknown protein 32 from 2D-page of liver tissue
Gene Symbols PBLD
Host Species Mouse
Immunogen Full length human recombinant protein of PBLD produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64081
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.