Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PBMC Mouse anti-Human, Unconjugated, Clone: 2E3, Abnova™

Catalog No. 89335166 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-335-166 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-335-166 Supplier Abnova Corporation Supplier No. MAB19482M01
Add to Cart
Add to Cart

Mouse Monoclonal Antibody.

PBMC Monoclonal antibody specifically detects PBMC in Human samples. It is validated for ELISA, WB-Re.

Specifications

Antigen PBMC
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2E3
Conjugate Unconjugated
Host Species Mouse
Immunogen PBMC
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.