Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PBOV1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986922
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6922 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6922 Supplier Bioss Supplier No. BS5939R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This intronless gene encodes a protein of unknown function. Its expression is up-regulated in some types of cancer, including prostate, breast, and bladder cancer. [provided by RefSeq, Aug 2011].

Specifications

Antigen PBOV1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene PBOV1
Gene Accession No. Q9GZY1
Gene Alias dJ171N11.2; PBOV1; prostate and breast cancer overexpressed 1; Prostate and breast cancer overexpressed gene 1 protein; Protein UROC28; UC28; UROC28
Gene Symbols PBOV1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human PBOV1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 59351
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.