Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pbp2B Rabbit anti-Bacillus species, Unconjugated, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma ABS2199100UL
Description
Pbp2B Polyclonal antibody specifically detects Pbp2B in Bacillus species samples. It is validated for Immunofluorescence, Western Blotting.
Specifications
| Pbp2B | |
| Polyclonal | |
| NP_389399 | |
| PbpB | |
| His-tagged full-length recombinant Penicillin-binding protein 2B from Bacillus subtilis, strain 168CA. | |
| Unpurified | |
| Signaling | |
| This rabbit polyclonal antibody specifically detects Penicillin-binding protein 2B (Pbp2B) in Bacillus subtilis. | |
| Stable for 1 year at -20°C from date of receipt. |
| Immunofluorescence, Western Blot | |
| Unconjugated | |
| Penicillin-binding protein 2B;PBP-2B;Penicillin-binding protein B | |
| Rabbit | |
| ∼80 kDa observed; 79.32 kDa calculated. Uncharacterized bands may be observed in some lysate(s). | |
| 100 μL | |
| Primary | |
| Bacillus species | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction