Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PBR Antibody (23G2), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | PBR |
---|---|
Clone | 23G2 |
Dilution | Western Blot 1:500-1:5000, Flow Cytometry, ELISA, Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:20-1:200 |
Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation |
Classification | Monoclonal |
Description
PBR Monoclonal antibody specifically detects PBR in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation, ImmunocytochemistrySpecifications
PBR | |
Western Blot 1:500-1:5000, Flow Cytometry, ELISA, Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:20-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse | |
benzodiazepine peripheral binding site, BZRPDBI, IBP, MBRPBS, mDRC, Mitochondrial benzodiazepine receptor, PBRBPBS, PBRS, peripheral benzodiazepine receptor, Peripheral-type benzodiazepine receptor, pk18, PKBSbenzodiazapine receptor (peripheral), PTBR, translocator protein, translocator protein (18kDa) | |
A synthesized peptide derived from Human PBR [UniProt P30536] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
23G2 | |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Cancer | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
706 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title