Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PBRM1 Mouse anti-Human, Polyclonal, Abnova™
Mouse polyclonal antibody raised against a full-length human PBRM1 protein.
Supplier: Abnova Corporation H00055193B01P
Specifications
PBRM1 | |
Polyclonal | |
polybromo 1 | |
PBRM1 | |
BAF180, MGC156155, MGC156156, PB1 | |
Mouse | |
Affinity chromatography | |
RUO | |
55193 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
Western Blot | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
BC015323 | |
PBRM1 | |
PBRM1 (AAH15323, 1 a.a. ∽ 306 a.a) full-length human protein. | |
50 μg | |
Primary | |
Human | |
Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction