Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PBRM1 Recombinant Rabbit Monoclonal Antibody (9A8C4)
GREENER_CHOICE

Catalog No. PIMA537814
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA537814 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA537814 Supplier Invitrogen™ Supplier No. MA537814
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive Samples: HeLa, NIH/3T3, Jurkat, Rat brain Immunogen sequence: VLEAREPGSG RRLCDLFMVK PSKKDYPDYY KIILEPMDLK IIEHNIRNDK YAGEEGMIED MKLMFRNARH YNEEGSQVYN DAHILEKLLK EKRKELGPLP DDDDMASPKL KLSRKSGISP KKSKYMTPMQ QKLNEVYEAV KNYTDKRGRR L.

The PBRM1 gene encodes a subunit of the PBAF (Polybromo-associated BAF) chromatin remodeling complex, playing a crucial role in the regulation of chromatin structure and transcription. PBRM1, also known as BAF180, contains bromodomains that recognize acetylated lysine residues on histone tails, facilitating chromatin remodeling necessary for effective transcriptional regulation. This gene is frequently mutated in renal cell carcinoma, particularly clear cell renal cell carcinoma (ccRCC), where such mutations are implicated in the pathogenesis of the disease. PBRM1 acts as a tumor suppressor, and its loss of function can lead to deregulation of gene expression, contributing to oncogenic processes. It is involved in pathways that govern cell cycle regulation and DNA damage response, reflecting its importance in maintaining genomic stability. Ongoing research focuses on understanding PBRM1's interactions within chromatin remodeling complexes and its potential as a therapeutic target in cancer treatment.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PBRM1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone 9A8C4
Concentration 1.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Pbrm1
Gene Accession No. Q86U86, Q8BSQ9
Gene Alias 2310032M22Rik; 2610016F04Rik; AI507524; BAF180; BRG1-associated factor 180; glutamate receptor interacting protein 2; hPB1; LOC506557 protein; PB1; PBRM1; polybromo 1; polybromo-1D; Protein polybromo-1; RGD1565549
Gene Symbols Pbrm1
Host Species Rabbit
Immunogen Recombinant protein (or fragment)
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 306254, 55193, 66923
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.