Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PBS, 10X Solution, Molecular Biology Grade

Catalog No. aaj75889k2
Click to view available options
Quantity:
100 mL
1 L
5 L

For biological research. Affymetrix/USB™ PBS, 10X Solution, Molecular Biology Grade is 0.2μm filtered and dispensed into durable, square nalgene bottles and upright carton.

  • Consists of 80.6mM sodium phosphate, 19.4mM potassium phosphate, 27mM KCl and 1.37M NaCl in high purity dH2O
  • Filtered and dispensed into durable, square Nalgene bottles and upright carton with convenient dispensing spout
  • Dilute to 1X with high purity dH2O for a solution consisting of 10mM phosphate
TRUSTED_SUSTAINABILITY

Specifications

pH 7.4
Physical Form Solution
Quantity 1 L
Formulation 80.6 mM sodium phosphate, 19.4 mM potassium phosphate, 27 mM KCl and 1.37 M NaCl in high purity dH2O

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.