Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PBX1 Antibody (OTI1C11), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273265MFV450
Description
PBX1 Monoclonal antibody specifically detects PBX1 in Human, Mouse, Rat samples. It is validated for Flow CytometrySpecifications
PBX1 | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Mouse | |
Immunogen affinity purified | |
RUO | |
5087 | |
Store at 4°C in the dark. | |
IgG1 |
Flow Cytometry | |
OTI1C11 | |
Flow Cytometry | |
DKFZp686B09108, Homeobox protein PBX1, Homeobox protein PRL, MGC126627, Pre-B cell leukemia transcription factor-1, pre-B-cell leukemia homeobox 1, pre-B-cell leukemia transcription factor 1, PRL | |
Full length human recombinant protein of human PBX1(NP_002576) produced in HEK293 cell. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction