Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PBX1 Monoclonal Antibody (OTI1C11)
GREENER_CHOICE

Catalog No. PIMA525580
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA525580 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA525580 Supplier Invitrogen™ Supplier No. MA525580
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PBX1 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow

This gene encodes a nuclear protein that belongs to the PBX homeobox family of transcriptional factors. Studies in mice suggest that this gene may be involved in the regulation of osteogenesis, and required for skeletal patterning and programming. A chromosomal translocation, t(1; 19) involving this gene and TCF3/E2A gene, is associated with pre-B-cell acute lymphoblastic leukemia. The resulting fusion protein, in which the DNA binding domain of E2A is replaced by the DNA binding domain of this protein, transforms cells by constitutively activating transcription of genes regulated by the PBX protein family. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PBX1
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone OTI1C11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PBX1
Gene Accession No. P40424
Gene Alias 2310056B04Rik; D230003C07Rik; DKFZp686B09108; Homeobox protein PBX1; homeobox protein PRL; MGC126627; OTTHUMP00000032205; PBX homeobox 1; Pbx1; Pbx-1; pre B cell leukemia homeobox 1; pre-B-cell leukemia homeobox 1; pre-B-cell leukemia transcription factor 1; PRL
Gene Symbols PBX1
Host Species Mouse
Immunogen Full length human recombinant protein of PBX1 produced in HEK293 cell.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5087
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.