Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PBX1 Mouse anti-Human, Clone: OTI1C11, liquid, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene TA503065

Description
This gene encodes a nuclear protein that belongs to the PBX homeobox family of transcriptional factors. Studies in mice suggest that this gene may be involved in the regulation of osteogenesis, and required for skeletal patterning and programming. A chromosomal translocation, tinvolving this gene and TCF3/E2A gene, is associated with pre-B-cell acute lymphoblastic leukemia. The resulting fusion protein, in which the DNA binding domain of E2A is replaced by the DNA binding domain of this protein, transforms cells by constitutively activating transcription of genes regulated by the PBX protein family. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.Specifications
PBX1 | |
Monoclonal | |
0.29 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P40424 | |
PBX1 | |
Full length human recombit protein of human PBX1 produced in HEK293 cell. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
OTI1C11 | |
Unconjugated | |
PBX1 | |
CAKUHED | |
Mouse | |
Affinity Chromatography | |
RUO | |
5087 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction