Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PBX1 Polyclonal Antibody

PBX1 Polyclonal Antibody
Supplier: Thermo Scientific PA517223
Description
PBX1 Polyclonal Antibody for Western Blot, ICC/IF, IP, ChIP
Specifications
| PBX1 | |
| Polyclonal | |
| Unconjugated | |
| PBX1 | |
| 2310056B04Rik; D230003C07Rik; DKFZp686B09108; Homeobox protein PBX1; homeobox protein PRL; MGC126627; OTTHUMP00000032205; PBX homeobox 1; Pbx1; Pbx-1; pre B cell leukemia homeobox 1; pre-B-cell leukemia homeobox 1; pre-B-cell leukemia transcription factor 1; PRL | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 18514, 5087 | |
| -20°C | |
| Liquid |
| ChIP Assay, Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 19 μg/mL | |
| 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5 | |
| P40424, P41778 | |
| PBX1 | |
| Synthetic peptide corresponding to amino acids near the amino terminus of human Pbx1. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction