Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PBX2 Antibody (PCRP-PBX2-1C4) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314127
This item is not returnable.
View return policy
Description
PBX2 Monoclonal antibody specifically detects PBX2 in Human samples. It is validated for Flow Cytometry, ELISA, Immunofluorescence, Immunoprecipitation, Protein ArraySpecifications
PBX2 | |
Monoclonal | |
1 mg/mL | |
Flow Cytometry 1 to 2 μg /million cells, ELISA, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL, Immunoprecipitation 1 to 2 μg per 100-500 μg protein, Protein Array | |
G17Homeobox protein PBX2, HOX12homeobox 12, PBX2MHC, pre-B-cell leukemia homeobox 2, pre-B-cell leukemia transcription factor 2, Protein G17 | |
Recombinant full-length human PBX2 protein (Uniprot: P40425) | |
100 μg | |
Primary | |
Human | |
Purified |
Flow Cytometry, ELISA, Immunofluorescence, Immunoprecipitation, Peptide Array | |
PCRP-PBX2-1C4 | |
Unconjugated | |
10mM PBS without Preservative | |
Mouse | |
Protein A or G purified | |
RUO | |
5089 | |
Store at -20 to -80°C. Avoid freeze-thaw cycles. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction