Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PBX2 Antibody (PCRP-PBX2-1C4), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314127MFV500
Description
PBX2 Monoclonal antibody specifically detects PBX2 in Human samples. It is validated for Flow CytometrySpecifications
PBX2 | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
5089 | |
Store at 4C in the dark. | |
IgG2b |
Flow Cytometry | |
PCRP-PBX2-1C4 | |
Flow Cytometry | |
G17Homeobox protein PBX2, HOX12homeobox 12, PBX2MHC, pre-B-cell leukemia homeobox 2, pre-B-cell leukemia transcription factor 2, Protein G17 | |
Recombinant full-length human PBX2 protein (Uniprot: P40425) | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction