Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PBX2 Mouse anti-Human, Clone: PCRP-PBX2-1C4, Novus Biologicals™
SDP

Catalog No. p-7244814 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB177833 20 μg
NB177832 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB177833 Supplier Novus Biologicals Supplier No. NBP31393820UG
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

PBX2 Monoclonal antibody specifically detects PBX2 in Human samples. It is validated for Flow Cytometry, ELISA, Immunofluorescence, Immunoprecipitation, Protein Array

Specifications

Antigen PBX2
Applications Flow Cytometry, ELISA, Immunofluorescence, Immunoprecipitation, Peptide Array
Classification Monoclonal
Clone PCRP-PBX2-1C4
Concentration 0.2 mg/ml
Conjugate Unconjugated
Dilution Flow Cytometry 1-2 ug/million cells, ELISA, Immunofluorescence 1-2 ug/ml, Immunoprecipitation 1-2 ug/100-500 ug protein, Protein Array
Formulation 10 mM PBS with 0.05% BSA
Gene Accession No. P40425
Gene Alias G17Homeobox protein PBX2, HOX12homeobox 12, PBX2MHC, pre-B-cell leukemia homeobox 2, pre-B-cell leukemia transcription factor 2, Protein G17
Host Species Mouse
Immunogen Recombinant full-length human PBX2 protein (Uniprot: P40425)
Purification Method Protein A or G purified
Quantity 20 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5089
Target Species Human
Content And Storage Store at 4°C.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.