Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PBX2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541437
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541437 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541437 Supplier Invitrogen™ Supplier No. PA541437
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: PGGYGDNLGG GQMYSPREMR ANGSWQEAVT PSSVTSPTEG PGSVHSDTSN Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.

PBX2 is a ubiquitously expressed member of the TALE/PBX homeobox family. PBX2 gene was identified by its similarity to a homeobox gene which is involved in t(1;19) translocation in acute pre-B-cell leukemias. PBX2 is a transcriptional activator which binds to the TLX1 promoter. This gene encodes a ubiquitously expressed member of the TALE/PBX homeobox family. It was identified by its similarity to a homeobox gene which is involved in t(1;19) translocation in acute pre-B-cell leukemias. This protein is a transcriptional activator which binds to the TLX1 promoter. The gene is located within the major histocompatibility complex (MHC) on chromosome 6.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PBX2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene PBX2
Gene Accession No. P40425
Gene Alias AU043397; G17; homeobox 12; homeobox protein PBX2; HOX12; PBX homeobox 2; Pbx2; PBX2MHC; PBX2P1; pbxy; pbxy homeodomain protein; pre B cell leukemia homeobox 2; pre B-cell leukemia transcription factor 2; pre-B-cell leukemia homeobox 2; pre-B-cell leukemia homeobox 2 pseudogene 1; pre-B-cell leukemia homeobox 3; pre-B-cell leukemia transcription factor 2; pre-B-cell leukemia transcription factor 2; pre-B-cell leukemia homeobox 2; pre-B-cell leukemia transcription factor y; Protein G17; wu:fb73g03; XXbac-BPG300A18.13
Gene Symbols PBX2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human PBX2 (aa 381-430).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5089
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.