Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PBX2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563964
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563964 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563964 Supplier Invitrogen™ Supplier No. PA563964
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GDSYSASQVE SLRHSMGPGG YGDNLGGGQM YSPREMRANG SW Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 86%.

PBX2 is a ubiquitously expressed member of the TALE/PBX homeobox family. PBX2 gene was identified by its similarity to a homeobox gene which is involved in t(1;19) translocation in acute pre-B-cell leukemias. PBX2 is a transcriptional activator which binds to the TLX1 promoter. This gene encodes a ubiquitously expressed member of the TALE/PBX homeobox family. It was identified by its similarity to a homeobox gene which is involved in t(1;19) translocation in acute pre-B-cell leukemias. This protein is a transcriptional activator which binds to the TLX1 promoter. The gene is located within the major histocompatibility complex (MHC) on chromosome 6.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PBX2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PBX2
Gene Accession No. P40425
Gene Alias AU043397; G17; homeobox 12; homeobox protein PBX2; HOX12; PBX homeobox 2; Pbx2; PBX2MHC; PBX2P1; pbxy; pbxy homeodomain protein; pre B cell leukemia homeobox 2; pre B-cell leukemia transcription factor 2; pre-B-cell leukemia homeobox 2; pre-B-cell leukemia homeobox 2 pseudogene 1; pre-B-cell leukemia homeobox 3; pre-B-cell leukemia transcription factor 2; pre-B-cell leukemia transcription factor 2; pre-B-cell leukemia homeobox 2; pre-B-cell leukemia transcription factor y; Protein G17; wu:fb73g03; XXbac-BPG300A18.13
Gene Symbols PBX2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PBX2. Recombinant protein control fragment (Product #RP-100032).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5089
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.