Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PBX2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587730
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587730 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587730 Supplier Invitrogen™ Supplier No. PA587730
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AVSVTQGGHS RTSSPTPPSS AGSGGSFNLS GSGDMFLGMP GLNGDSYSAS QVESLRHSMG PGGYGDNLGG GQMYSPREMR ANGSWQEAVT PSSVTSPTEG PGSVHSDTSN; Positive Samples: Mouse liver, Rat liver; Cellular Location: Nucleus.

PBX2 is a ubiquitously expressed member of the TALE/PBX homeobox family. PBX2 gene was identified by its similarity to a homeobox gene which is involved in t(1;19) translocation in acute pre-B-cell leukemias. PBX2 is a transcriptional activator which binds to the TLX1 promoter. This gene encodes a ubiquitously expressed member of the TALE/PBX homeobox family. It was identified by its similarity to a homeobox gene which is involved in t(1;19) translocation in acute pre-B-cell leukemias. This protein is a transcriptional activator which binds to the TLX1 promoter. The gene is located within the major histocompatibility complex (MHC) on chromosome 6.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PBX2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.83 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PBX2
Gene Accession No. O35984
Gene Alias AU043397; G17; homeobox 12; homeobox protein PBX2; HOX12; PBX homeobox 2; Pbx2; PBX2MHC; PBX2P1; pbxy; pbxy homeodomain protein; pre B cell leukemia homeobox 2; pre B-cell leukemia transcription factor 2; pre-B-cell leukemia homeobox 2; pre-B-cell leukemia homeobox 2 pseudogene 1; pre-B-cell leukemia homeobox 3; pre-B-cell leukemia transcription factor 2; pre-B-cell leukemia transcription factor 2; pre-B-cell leukemia homeobox 2; pre-B-cell leukemia transcription factor y; Protein G17; wu:fb73g03; XXbac-BPG300A18.13
Gene Symbols PBX2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 321-430 of human PBX2 (NP_0025772).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18515, 406164
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.