Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PBX3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110075
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110075 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110075 Supplier Invitrogen™ Supplier No. PA5110075
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MDDQSRMLQT LAGVNLAGHS VQGGMALPPP PHGHEGADGD GRKQDIGDIL HQIMTITDQS LDEAQAKKHA LNCHRMKPAL FSVLCEIKEK TGLSIRGAQE.

PBX3 is a transcriptional activator that binds the sequence 5'-ATCAATCAA-3'.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PBX3
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.55 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene PBX3
Gene Accession No. O35317, P40426
Gene Alias homeobox protein PBX3; PBX homeobox 3; Pbx3; pre B cell leukemia homeobox 3; pre B-cell leukemia transcription factor 3; pre-B-cell leukemia homeobox 3; pre-B-cell leukemia transcription factor 3; Unknown (protein for MGC:134053)
Gene Symbols PBX3
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human PBX3 (NP_0061861).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18516, 5090
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.