Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PBX4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA568279
Description
PBX4 Polyclonal Antibody for Western Blot
PBX4 encodes a member of the pre-B cell leukemia transcription factor family. These proteins are homeobox proteins that play critical roles in embryonic development and cellular differentiation both as Hox cofactors and through Hox-independent pathways. The encoded protein contains a homeobox DNA-binding domain, but specific functions of the protein have not been determined. Alternatively spliced transcript variants have been observed for this gene.
Specifications
PBX4 | |
Polyclonal | |
Unconjugated | |
PBX4 | |
2410015M21Rik; AI429113; AW045266; Edg4; endothelial differentiation, lysophosphatidic acid G-protein-coupled receptor 4; homeobox protein PBX4; PBX homeobox 4; PBX4; pre B cell leukemia homeobox 4; pre-B-cell leukemia homeobox 4; pre-B-cell leukemia transcription factor 4 | |
Rabbit | |
Affinity chromatography | |
RUO | |
80714 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q9BYU1 | |
PBX4 | |
Synthesized peptide derived from human PBX4. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction