Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PBX4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568872
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568872 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568872 Supplier Invitrogen™ Supplier No. PA568872
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Mouse: 100%; Rat: 92%.

PBX4 encodes a member of the pre-B cell leukemia transcription factor family. These proteins are homeobox proteins that play critical roles in embryonic development and cellular differentiation both as Hox cofactors and through Hox-independent pathways. The encoded protein contains a homeobox DNA-binding domain, but specific functions of the protein have not been determined. Alternatively spliced transcript variants have been observed for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PBX4
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene PBX4
Gene Accession No. Q99NE9
Gene Alias 2410015M21Rik; AI429113; AW045266; Edg4; endothelial differentiation, lysophosphatidic acid G-protein-coupled receptor 4; homeobox protein PBX4; PBX homeobox 4; PBX4; pre B cell leukemia homeobox 4; pre-B-cell leukemia homeobox 4; pre-B-cell leukemia transcription factor 4
Gene Symbols PBX4
Host Species Rabbit
Immunogen synthetic peptide corresponding to a region of mouse PBX4 (aa 1-50).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80720
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.