Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PBX4 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982116
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2116 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2116 Supplier Bioss Supplier No. BS12298R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PBX4 encodes a member of the pre-B cell leukemia transcription factor family. These proteins are homeobox proteins that play critical roles in embryonic development and cellular differentiation both as Hox cofactors and through Hox-independent pathways. The encoded protein contains a homeobox DNA-binding domain, but specific functions of the protein have not been determined. Alternatively spliced transcript variants have been observed for this gene.

Specifications

Antigen PBX4
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene PBX4
Gene Accession No. Q9BYU1
Gene Alias 2410015M21Rik; AI429113; AW045266; Edg4; endothelial differentiation, lysophosphatidic acid G-protein-coupled receptor 4; Homeobox protein PBX4; PBX homeobox 4; Pbx4; pre B cell leukemia homeobox 4; pre-B-cell leukemia homeobox 4; Pre-B-cell leukemia transcription factor 4
Gene Symbols PBX4
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human PBX4.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80714
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.