Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PBXIP1 Antibody, Janelia Fluor™ 549, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP305982JF549
Description
PBXIP1 Polyclonal specifically detects PBXIP1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
PBXIP1 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
Hematopoietic PBX-interacting protein, HPIPpre-B-cell leukemia transcription factor interacting protein 1, pre-B-cell leukemia homeobox interacting protein 1, pre-B-cell leukemia transcription factor-interacting protein 1 | |
Produced in rabbits immunized with E. coli-derived Human PBXIP1 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Janelia Fluor 549 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
57326 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction