Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PBXIP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552366
Description
Immunogen sequence: SLPVETLGPA SRMDPESERA LQAPHSPSKT DGKELAGTMD GEGTLFQTES PQSGSILTEE TEVKGTLEGD VCGVEPPGPG DTVVQGDLQE TTVVTGLGPD TQDLEGQSPP QSLPSTPKAA WIR Highest antigen sequence identity to the following orthologs: Mouse - 62%, Rat - 62%.
Regulator of pre-B-cell leukemia transcription factors (BPXs) function. Inhibits the binding of PBX1-HOX complex to DNA and blocks the transcriptional activity of E2A-PBX1. Tethers estrogen receptor-alpha (ESR1) to microtubules and allows them to influence estrogen receptors-alpha signaling.
Specifications
PBXIP1 | |
Polyclonal | |
Unconjugated | |
PBXIP1 | |
4732463H20Rik; hematopoietic PBX-interacting protein; HPIP; PBX homeobox interacting protein 1; PBXIP1; pre B cell leukemia transcription factor interacting protein 1; pre-B-cell leukemia homeobox interacting protein 1; pre-B-cell leukemia transcription factor interacting protein 1; pre-B-cell leukemia transcription factor-interacting protein 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57326 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96AQ6 | |
PBXIP1 | |
Recombinant protein corresponding to Human PBXIP1. Recombinant protein control fragment (Product #RP-89383). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction