Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PBXIP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598792
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598792 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598792 Supplier Invitrogen™ Supplier No. PA598792
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PBXIP1 Polyclonal Antibody for Western Blot, ELISA

Regulator of pre-B-cell leukemia transcription factors (BPXs) function. Inhibits the binding of PBX1-HOX complex to DNA and blocks the transcriptional activity of E2A-PBX1. Tethers estrogen receptor-alpha (ESR1) to microtubules and allows them to influence estrogen receptors-alpha signaling.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PBXIP1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene PBXIP1
Gene Accession No. Q96AQ6
Gene Alias 4732463H20Rik; hematopoietic PBX-interacting protein; HPIP; PBX homeobox interacting protein 1; PBXIP1; pre B cell leukemia transcription factor interacting protein 1; pre-B-cell leukemia homeobox interacting protein 1; pre-B-cell leukemia transcription factor interacting protein 1; pre-B-cell leukemia transcription factor-interacting protein 1
Gene Symbols PBXIP1
Host Species Rabbit
Immunogen Recombinant Human Pre-B-cell leukemia transcription factor-interacting protein 1 protein (361-600AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57326
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.