Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PBXIP1 Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501727715 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-7715 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-7715 Supplier Proteintech Group Inc Supplier No. 121021AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The predicted PBXIP1 protein has a calculated molecular mass of 81 kDa. This polyclonal antibody raised against 1-300 aa of human PBXIP1 recognizes endogenous PBXIP1 with an apparent molecular weight of 95-100 kDa. The slow migration of PBXIP1 possibly re

Regulator of pre-B-cell leukemia transcription factors (BPXs) function. Inhibits the binding of PBX1-HOX complex to DNA and blocks the transcriptional activity of E2A-PBX1. Tethers estrogen receptor-alpha (ESR1) to microtubules and allows them to influence estrogen receptors-alpha signaling.

Specifications

Antigen PBXIP1
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene PBXIP1
Gene Accession No. Q96AQ6
Gene Alias HPIP, PBXIP1
Gene Symbols PBXIP1
Host Species Rabbit
Immunogen PBXIP1 Fusion Protein Ag2746
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57326
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.