Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PC1/3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA579787
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA579787 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA579787 Supplier Invitrogen™ Supplier No. PA579787
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: Rat Liver Tissue, Rat Thymus Tissue, A549 Cell, HELA Cell, COLO320 Cell, PANC Cell. IHC: rat intestine tissue.

PCSK1 (Proprotein Convertase, Subtilisin/Kexin-Type, 1), also known as PC1 or NEC1, is an enzyme that in humans is encoded by the PCSK1 gene. Proprotein convertase-1 is a neuroendocrine convertase that belongs to a family of subtilisin-like serine endoproteases that process large precursor proteins into mature bioactive products. By in situ hybridization, Seidah et al. (1991) mapped the NEC1 gene to human chromosome 5q15-q21 and mouse chromosome 13. Ohagi et al. (1996) noted that PC1 initiates the sequential processing of proinsulin to insulin by cleaving the proinsulin molecule on the C-terminal side of the dibasic peptide, arg31-arg32, joining the B-chain and C-peptide. By observing the phenotypic features in patients with PC1 mutations, Jackson et al. (2003) concluded that human intestinal absorptive function is dependent on PC1 activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PC1/3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 5mg BSA and 0.05mg sodium azide, 0.05mg thimerosal
Gene PCSK1
Gene Accession No. P28840, P29120, P63239
Gene Alias Att-1; BDP; BMIQ12; furin homolog; NEC 1; Nec1; Nec-1; Neuroendocrine convertase 1; PC1; PC3; PCSK1; Phpp-1; Prohormone convertase 1; prohormone convertase 1/3; prohormone convertase 3; propeptide-processing protease; Proprotein convertase 1; proprotein convertase subtilisin/kexin type 1; Protein convertase subtilisin / kexin type I; Protein convertase subtilisin / kexin, type I; SPC3
Gene Symbols PCSK1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence at the C-terminus of human PC1/3 (726-740aa DVFYNTKPYKHRDDR).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18548, 25204, 5122
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.