Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PC4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117086
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117086 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5117086 Supplier Invitrogen™ Supplier No. PA5117086
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: HL-60, BT-474, SW620, Jurkat, Mouse brain Immunogen sequence: MPKSKELVSS SSSGSDSDSE VDKKLKRKKQ VAPEKPVKKQ KTGETSRALS SSKQSSSSRD DNMFQIGKMR YVSVRDFKGK VLIDIREYWM DPEGEMKPGR KGISLNPEQW SQLKEQISDI DDAVRKL.

PC4 (activated RNA polymerase II transcription cofactor 4) is a transcriptional coactivator, possessing the ability to suppress promoter-driven as well as nonspecific transcription via its DNA binding activity. The repressive activity of PC4 on promoter-driven transcription is alleviated by transcription factor TFIIH. TFIIH protects promoters from PC4-mediated repression by relieving the topological constraint imposed by PC4 through the ERCC3 helicase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PC4
Applications ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.11 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Sub1
Gene Accession No. P11031, P53999
Gene Alias activated RNA polymerase II transcription cofactor 4; activated RNA polymerase II transcriptional coactivator p15; AI842364; hypothetical protein LOC553561; p14; P15; P9; Pc4; positive cofactor 4; pR-ET2 encoded oncodevelopmental protein; RNA polymerase II transcriptional coactivator; Rpo2tc1; Single-stranded DNA-binding protein p9; Sub1; SUB1 homolog; SUB1 homolog (S. cerevisiae); SUB1 homolog a; SUB1 homolog, transcriptional regulator; SUB1 homolog, transcriptional regulator a; SUB1 regulator of transcription; SUB1 regulator of transcription a; sub1a; TIS7; zgc:109973
Gene Symbols Sub1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-127 of human SUB1 (NP_0067043).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10923, 20024
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.