Learn More
PC4 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$167.95 - $630.77
Specifications
| Antigen | PC4 |
|---|---|
| Applications | Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Liquid |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-157-1724
|
Bethyl Laboratories, Inc
IHC00231T |
10 μL |
Each for $167.95
|
|
|||||
50-157-1723
|
Bethyl Laboratories, Inc
IHC00231 |
100 μL |
Each for $630.77
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Rat, Chicken, Turkey, Bovine, Dog, Horse, Rabbit, Guinea pig_10141, Pig, Panda, Orangutan,
PC4 (activated RNA polymerase II transcription cofactor 4) is a transcriptional coactivator, possessing the ability to suppress promoter-driven as well as nonspecific transcription via its DNA binding activity. The repressive activity of PC4 on promoter-driven transcription is alleviated by transcription factor TFIIH. TFIIH protects promoters from PC4-mediated repression by relieving the topological constraint imposed by PC4 through the ERCC3 helicase activity.Specifications
| PC4 | |
| Polyclonal | |
| Liquid | |
| RUO | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| PC4, TIS7 | |
| Between 77 and 127 | |
| Primary | |
| 4° C | |
| SUB1 |
| Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| P11031, P53999 | |
| 10923, 20024 | |
| IgG | |
| Antigen affinity chromatography | |
| Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.