Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PC6 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP157908
Description
PC6 Polyclonal specifically detects PC6 in Human samples. It is validated for Western Blot.Specifications
PC6 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
EC 3.4.21, EC 3.4.21.-, EC 3.4.21.61, EC 3.4.21.75, FLJ11149, FLJ16215, hPC6, PC5PC6A, PC6prohormone convertase 5, Proprotein convertase 5, Proprotein convertase 6, proprotein convertase PC5, proprotein convertase subtilisin/kexin type 5, protease PC6, SPC6, Subtilisin/kexin-like protease PC5 | |
Rabbit | |
Affinity purified | |
RUO | |
5125 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
Q92824 | |
PCSK5 | |
Synthetic peptides corresponding to PCSK5(proprotein convertase subtilisin/kexin type 5) The peptide sequence was selected from the middle region of PCSK5. Peptide sequence CAGAGADGCINCTEGYFMEDGRCVQSCSISYYFDHSSENGYKSCKKCDIS. | |
100 μL | |
Primary | |
Expected identity based on immunogen sequence: Chicken: 76%. | |
Human, Mouse, Rat, Bovine, Canine, Equine, Rabbit | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction