Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Promega pCAT3™ Vectors
Click to view available options
Vector:
pCAT3-control vector
pCAT3-enhancer vector
pCAT3-promoter vector
Description
- Reporter Vectors that Express Chloramphenicol Acetyltransferase (CAT)
- Offer reduced background CAT expression
- Increased expression with more efficient poly(A) signal
- Can produce ssDNA for sequencing and mutagenesis
Specifications
Specifications
For Use With (Equipment) | CAT reporter system |
Content And Storage | -30°C to -10°C |
Quantity | 20 μg |
Vector | pCAT3-control vector |
For Use With (Application) | Quantitative analysis of factors that potentially regulate mammalian gene expression |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction