Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PCBD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566416
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566416 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566416 Supplier Invitrogen™ Supplier No. PA566416
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YNKVHITLSTH ECAGLSERDI NLASFIEQVA VSMT Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.

PCBD1 encodes pterin-4 alpha-carbinolamine dehydratase, an enzyme involved in phenylalanine hydroxylation. A deficiency of this enzyme leads to hyperphenylalaninemia. The enzyme regulates the homodimerization of the transcription factor hepatocyte nuclear factor 1 (HNF1).
TRUSTED_SUSTAINABILITY

Specifications

Antigen PCBD1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PCBD1
Gene Accession No. P61457
Gene Alias 4-alpha-hydroxy-tetrahydropterin dehydratase; 6-pyruvoyl-tetrahydropterin synthase/dimerization cofactor of hepatocyte nuclear factor 1 alpha (TCF1); DCOH; Dimerization cofactor of hepatocyte nuclear factor 1-alpha; dimerization cofactor of HNF1; dimerizing cofactor for HNF1; Pcbd; PCBD1; PCD; Phenylalanine hydroxylase-stimulating protein; PHS; pt; pterin 4 alpha carbinolamine dehydratase/dimerization cofactor of hepatocyte nuclear factor 1 alpha (TCF1) 1; pterin carbinolamine dehydratase; pterin-4 alpha-carbinolamine dehydratase 1; pterin-4 alpha-carbinolamine dehydratase/dimerization cofactor of hepatocyte nuclear factor 1 alpha; pterin-4-alpha-carbinolamine dehydratase; TCF1
Gene Symbols PCBD1
Host Species Rabbit
Immunogen Recombinant Human PCBD1. Recombinant protein control fragment (Product #RP-105203).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5092
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.