Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PCBP1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA565369
Description
Immunogen sequence: QHTITPLDLAK LNQVARQQSH FAMMHGG Highest antigen sequence identity to the following orthologs - mouse 96%, rat 57%.
This intronless gene is thought to have been generated by retrotransposition of a fully processed PCBP-2 mRNA. This gene and PCBP-2 have paralogues which are thought to have arisen as a result of duplication events of entire genes. The protein encoded by this gene appears to be multifunctional. It along with PCBP-2 and hnRNPK corresponds to the major cellular polydomains which may be involved in RNA binding. This encoded protein together with PCBP-2 also functions as translational coactivators of poliovirus RNA via a sequence-specific interaction with stem-loop IV of the IRES and promote poliovirus RNA replication by binding to its 5'-terminal cloverleaf structure. It has also been implicated in translational control of the 15-lipoxygenase mRNA, human Papillomavirus type 16 L2 mRNA, and hepatitis A virus RNA. The encoded protein is also suggested to play a part in formation of a sequence-specific alpha-globin mRNP complex which is associated with alpha-globin mRNA stability.
Specifications
| PCBP1 | |
| Polyclonal | |
| Unconjugated | |
| PCBP1 | |
| [a]CP-1; Alpha-CP1; alphaCP-1; epididymis secretory protein Li 85; HEL-S-85; Heterogeneous nuclear ribonucleoprotein E1; heterogenous nuclear ribonucleoprotein E1; heterogenous nuclear ribonucleoprotein X; hnRNP E1; hnRNP-E1; hnRNP-E1 protein; hnRNP-X; HNRPE1; HNRPX; Nucleic acid-binding protein SUB2.3; PCBP1; poly(rC) binding protein 1; poly(rC)-binding protein 1; RGD1561319; WBP17 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5093 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q15365 | |
| PCBP1 | |
| Recombinant Human PCBP1. Recombinant protein control fragment (Product #RP-104777). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction