Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PCBP1 Rabbit anti-Human, Mouse, Rat, Clone: 7M1K0, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | PCBP1 |
---|---|
Clone | 7M1K0 |
Dilution | Western Blot 1:500 - 1:2000 |
Applications | Western Blot |
Classification | Monoclonal |
Description
PCBP1 Monoclonal antibody specifically detects PCBP1 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
PCBP1 | |
Western Blot 1:500 - 1:2000 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
alpha-CP1, Heterogeneous nuclear ribonucleoprotein E1, heterogenous nuclear ribonucleoprotein E1, heterogenous nuclear ribonucleoprotein X, hnRNP E1, hnRNP-E1, hnRNP-X, HNRPE1, HNRPX, nucleic acid binding protein sub 2.3, Nucleic acid-binding protein SUB2.3, poly(rC) binding protein 1, poly(rC)-binding protein 1 | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human PCBP1 (Q15365). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
7M1K0 | |
Western Blot | |
Unconjugated | |
Rabbit | |
DNA replication Transcription Translation and Splicing | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
5093 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title